Merck
  • The role of mannitol as a nephroprotectant in patients receiving cisplatin therapy.
The role of mannitol as a nephroprotectant in patients receiving cisplatin therapy.
The Annals of pharmacotherapy (2012-02-03)
Katherine P Morgan, Larry W Buie, Scott W Savage
ABSTRACT
To review the efficacy and safety of concomitant mannitol administration with cisplatin therapy to reduce the incidence of nephrotoxicity. A literature search was performed via MEDLINE, PubMed, and the Cochrane Library (1945-August 2011) using the terms mannitol, cisplatin, nephrotoxicity, and forced diuresis. Reference citations from the publications identified were also reviewed. The search was limited to studies conducted in humans. All studies in which mannitol was used for forced diuresis with cisplatin therapy were evaluated. Cisplatin therapy can lead to transient and permanent renal impairment. Molecular and histologic changes occur in the renal tubules, which contribute to nephrotoxicity. The adverse effect profile of cisplatin is well documented, but the prevention strategies to alleviate renal impairment due to treatment are less understood. Mannitol plus hydration has been used for several years to alleviate toxicity associated with cisplatin therapy. However, the data for mannitol administration have not been convincing. When the use of mannitol and hydration is compared directly to hydration alone, mannitol shows no benefit. In some patients, not only was mannitol not protective, its administration was associated with worsening renal function. Although mannitol plus hydration is used to decrease cisplatin-induced nephrotoxicity, there are no compelling data that the addition of mannitol is more nephroprotective than the use of hydration alone. Appropriate hydration remains the most reasonable strategy to reduce the incidence of cisplatin-induced nephrotoxicity.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination