Merck
Niacin: another look at an underutilized lipid-lowering medication.
Nature reviews. Endocrinology (2012-02-22)
Julia C Creider, Robert A Hegele, Tisha R Joy
ABSTRACT
Niacin, or water-soluble vitamin B(3), when given at pharmacologic doses, is a powerful lipid-altering agent. This drug, which lowers the levels of atherogenic, apolipoprotein-B-containing lipoproteins, is one of few medications that can raise the levels of atheroprotective HDL cholesterol. Niacin also has beneficial effects on other cardiovascular risk factors, including lipoprotein(a), C-reactive protein, platelet-activating factor acetylhydrolase, plasminogen activator inhibitor 1 and fibrinogen. Many clinical trials have confirmed the lipid effects of niacin treatment; however, its effects on cardiovascular outcomes have been called into question owing to the AIM-HIGH trial, which showed no benefit of niacin therapy on cardiovascular endpoints. Furthermore, use of niacin has historically been limited by tolerability issues. In addition to flushing, worsened hyperglycaemia among patients with diabetes mellitus has also been a concern with niacin therapy. This article reviews the utility of niacin including its mechanism of action, clinical trial data regarding cardiovascular outcomes, adverse effect profile and strategies to address these effects and improve compliance.
Product Description
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
USP
Niacin, United States Pharmacopeia (USP) Reference Standard
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard