Merck
  • Cellular defense mechanisms against lead toxicity in the vascular system.
Cellular defense mechanisms against lead toxicity in the vascular system.
Biological & pharmaceutical bulletin (2012-11-06)
Yasuhiro Shinkai, Toshiyuki Kaji
ABSTRACT
Lead is a toxic heavy metal that can cause a range of health problems. In this context, the vascular system is a particular target of the deleterious effects of lead. Lead exerts its toxicity through substitution of other divalent cations such as calcium and zinc, resulting in disruption of homeostasis. Based on the evidence that lead up-regulates endoplasmic reticulum (ER) chaperone glucose-regulated protein 78 (GRP78) and/or antioxidant proteins such as hemeoxygenase-1, it is believed that the heavy metal is able to induce ER and/or oxidative stress in cells. These events also suggest that the unfolded protein response (UPR) system and the antioxidant defense system Kelch-like ECH-associated protein 1-nuclear factor (NF)-E2-related factor 2 (Keap1-Nrf2) play a critical role in adaptive response to lead. In this review, we summarize recent progress in lead toxicity in terms of cellular defense systems, including stress proteins and transcription factors involved in the vascular system.
Product Description
Lead, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 5m coil, thickness 0.125mm, as rolled, 99.95%
Lead, rod, 100mm, diameter 19.0mm, 99.95%
Lead, foil, 300x300mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, not light tested, 300x300mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 50x50mm, thickness 0.03mm, 99.99+%
Lead, foil, 1m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Lead, wire reel, 2m, diameter 0.5mm, as drawn, 99.999%
Lead, foil, 100x100mm, thickness 0.5mm, as rolled, 99.95%
Lead, rod, 1000mm, diameter 12.7mm, 99.95%
Lead, foil, 50x50mm, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Lead, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 50x50mm, thickness 0.01mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 8mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 2m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, rod, 100mm, diameter 6.35mm, 99.95%
Lead, foil, 10mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 50x50mm, thickness 2.7mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.006mm, 99.99+%
Lead, foil, 6mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 15mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 1m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)