Merck
  • A newly discovered function of peroxisomes: involvement in biotin biosynthesis.
A newly discovered function of peroxisomes: involvement in biotin biosynthesis.
Plant signaling & behavior (2012-10-18)
Jun-ichi Maruyama, Shohei Yamaoka, Ichiro Matsuo, Nobuhiro Tsutsumi, Katsuhiko Kitamoto
ABSTRACT
In plants, peroxisomes are the organelles involved in various metabolic processes and physiological functions including β-oxidation, mobilization of seed storage lipids, photorespiration, and hormone biosynthesis. We have recently shown that, in fungi and plants, peroxisomes play a vital role in biosynthesis of biotin, an essential cofactor required for various carboxylation and decarboxylation reactions. In fungi, the mutants defective in peroxisomal protein import exhibit biotin auxotrophy. The fungal BioF protein, a 7-keto-8-aminopelargonic acid (KAPA) synthase catalyzing the conversion of pimeloyl-CoA to KAPA in biotin biosynthesis, contains the peroxisomal targeting sequence 1 (PTS1), and its peroxisomal targeting is required for biotin biosynthesis. In plants, biotin biosynthesis is essential for embryo development. We have shown that the peroxisomal targeting sequences of the BioF proteins are conserved throughout the plant kingdom, and the Arabidopsis thaliana BioF protein is indeed localized in peroxisomes. Our findings suggest that peroxisomal localization of the BioF protein is evolutionarily conserved among eukaryotes, and required for biotin biosynthesis and plant growth and development.
Product Description
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Biotin, European Pharmacopoeia (EP) Reference Standard
USP
Biotin, United States Pharmacopeia (USP) Reference Standard
SAFC
BIOTIN
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material