Merck
  • A modular, energy-based approach to the development of nickel containing molecular electrocatalysts for hydrogen production and oxidation.
A modular, energy-based approach to the development of nickel containing molecular electrocatalysts for hydrogen production and oxidation.
Biochimica et biophysica acta (2013-01-15)
Wendy J Shaw, Monte L Helm, Daniel L DuBois
ABSTRACT
This review discusses the development of molecular electrocatalysts for H2 production and oxidation based on nickel. A modular approach is used in which the structure of the catalyst is divided into first, second, and outer coordination spheres. The first coordination sphere consists of the ligands bound directly to the metal center, and this coordination sphere can be used to control such factors as the presence or absence of vacant coordination sites, redox potentials, hydride donor abilities and other important thermodynamic parameters. The second coordination sphere includes functional groups such as pendent acids or bases that can interact with bound substrates such as H2 molecules and hydride ligands, but that do not form strong bonds with the metal center. These functional groups can play diverse roles such as assisting the heterolytic cleavage of H2, controlling intra- and intermolecular proton transfer reactions, and providing a physical pathway for coupling proton and electron transfer reactions. By controlling both the hydride donor ability of the catalysts using the first coordination sphere and the proton donor abilities of the functional groups in the second coordination sphere, catalysts can be designed that are biased toward H2 production, oxidation, or bidirectional (catalyzing both H2 oxidation and production). The outer coordination sphere is defined as that portion of the catalytic system that is beyond the second coordination sphere. This coordination sphere can assist in the delivery of protons and electrons to and from the catalytically active site, thereby adding another important avenue for controlling catalytic activity. Many features of these simple catalytic systems are good models for enzymes, and these simple systems provide insights into enzyme function and reactivity that may be difficult to probe in enzymes. This article is part of a Special Issue entitled: Metals in Bioenergetics and Biomimetics Systems.
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
Nickel, broken cathode squares, 500g, max. size 50mm, 99.8%
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Nickel, wire reel, 50m, diameter 1.0mm, as drawn, 99%
Nickel, rod, 100mm, diameter 2.0mm, annealed, 99%
Nickel, pellets, 100g, max. size 10mm, 99.99+%
Nickel, rod, 100mm, diameter 40mm, 99%
Nickel, rod, 500mm, diameter 12.7mm, 99.98%
Nickel, wire reel, 5m, diameter 0.5mm, as drawn, 99.99+%
Nickel, wire reel, 1m, diameter 2.0mm, hard, 99.98%
Nickel, foil, light tested, 100x100mm, thickness 0.015mm, 99.9%
Nickel, insulated wire, 50m, conductor diameter 0.025mm, insulation thickness 0.0035mm, polyimide insulation
Nickel, wire reel, 200m, diameter 0.125mm, annealed, 99.98%
Nickel, foil, light tested, 25x25mm, thickness 0.015mm, 99.9%
Nickel, foil, 25mm disks, thickness 0.05mm, as rolled, 99.98%
Nickel, rod, 200mm, diameter 3.0mm, 99%
Nickel, tube, 1000mm, outside diameter 2.4mm, inside diameter 2.248mm, wall thickness 0.076mm, hard, 99.5%
Nickel, foil, 15mm disks, thickness 0.125mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.5mm, as rolled, 100%
Nickel, tube, 100mm, outside diameter 1.8mm, inside diameter 1.6mm, wall thickness 0.1mm, hard, 99.5%
Nickel, foil, 15mm disks, thickness 0.075mm, as rolled, 99.98%
Nickel, rod, 500mm, diameter 3.0mm, 99%