Merck
Corneal crosslinking with riboflavin and ultraviolet A. I. Principles.
The ocular surface (2013-04-16)
Frederik Raiskup, Eberhard Spoerl
ABSTRACT
Changes in the biomechanical properties of the human cornea play an important role in the pathogenesis of corneal ectatic diseases. Biomechanical investigation shows significant differences between human ectatic corneas and normal corneas, including decreased stiffness and reduction of collagen crosslinks in the ectatic cornea. Induction of crosslinks is a well-established procedure in polymer chemistry to increase the elastic modulus of materials. Crosslinking (CXL) in connective tissue can occur during aging and as a side effect of diabetes mellitus. CXL has been used medically to increase stability and reduce the biodegradation of collagen-based biomaterials for bioprostheses. CXL of the cornea using riboflavin and UVA light with a wavelength of 370 nm and a dosage of 5.4 J/cm² is a new approach that increases the mechanical and biochemical stability of stromal tissue. This technique combines the principles of CXL (chemical and nonenzymatic) and the biochemical mechanisms of photo-oxidative CXL with riboflavin as a photosensitizer. In this review, the enrichment of riboflavin in the stroma by standard (epi-off) and transepithelial (epi-on) CXL is discussed. The theoretical and experimental measurements of the absorption of UV light explain the stronger CXL effect in the anterior stroma and its importance for the prevention of damage to the endothelial cells. UV devices are described. Changes of the physical properties after CXL, as well as the cellular changes, are discussed. From these basic investigations, treatment parameters for effective and safe CXL are identified.
Product Description
Supelco
Riboflavin (B2), analytical standard
Pricing and availability is not currently available.
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Pricing and availability is not currently available.
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.