Merck
  • Sodium acetate as a replacement for sodium bicarbonate in medical toxicology: a review.
Sodium acetate as a replacement for sodium bicarbonate in medical toxicology: a review.
Journal of medical toxicology : official journal of the American College of Medical Toxicology (2013-05-03)
Mark J Neavyn, Edward W Boyer, Steven B Bird, Kavita M Babu
ABSTRACT
Sodium bicarbonate is central to the treatment of many poisonings. When it was placed on the FDA drug shortage list in 2012, alternative treatment strategies to specific poisonings were considered. Many hospital pharmacies, poison centers, and medical toxicologists proposed sodium acetate as an adequate alternative, despite a paucity of data to support its use in medical toxicology. The intention of this review is to educate the clinician on the use of sodium acetate and to advise them on the potential adverse events when given in excess. We conducted a literature search focused on the pharmacology of sodium acetate, its use as a buffer in pathologic acidemia and dialysis baths, and potential adverse events associated with excess sodium acetate infusion. It appears safe to replace sodium bicarbonate infusion with sodium acetate on an equimolar basis. The metabolism of acetate, however, is more complex than bicarbonate. Future prospective studies will be needed to confirm the efficacy of sodium acetate in the treatment of the poisoned patient.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%