Merck
Dipyridamole and paracetamol overdose resulting in multi-organ failure.
Scottish medical journal (2013-08-21)
P S Cullis, D Watson, A Cameron, R F McKee
ABSTRACT
Dipyridamole intoxication is rare and few reports exist amongst the current literature. A case of dipyridamole and paracetamol overdose is described in a previously healthy 58-year-old woman, which resulted in multi-organ failure requiring dialysis, inotropic support, ventilation and extensive surgical intervention for small bowel ischaemia. This case highlights the dangers of an unusually large overdose of a commonly prescribed drug, and reviews current knowledge of dipyridamole intoxication.
Sigma-Aldrich
Dipyridamole, ≥98% (HPLC)
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Dipyridamole, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dipyridamole for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard