Merck
  • Carvone as a versatile chiral building block for total syntheses of heterocyclic sesquiterpenoids.
Carvone as a versatile chiral building block for total syntheses of heterocyclic sesquiterpenoids.
Natural product communications (2013-08-29)
Hisahiro Hagiwara
ABSTRACT
This review article is focused on the total syntheses of heterocyclic sesquiterpenoids starting from carvone since 1996.
Sigma-Aldrich
L-Carvone, natural, 99%, FG
Sigma-Aldrich
L-Carvone, ≥97%, FCC, FG
Supelco
(−)-Carvone, analytical standard
Sigma-Aldrich
(S)-(+)-Carvone, 96%
Supelco
(+)-Carvone, analytical standard
Sigma-Aldrich
(R)-(−)-Carvone, 98%
Sigma-Aldrich
D-Carvone, ≥96%, FG
Supelco
(+)-Carvone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland