Merck
[The specificity of silver nitrate staining of nucleolar organizers].
Tsitologiia (1989-01-01)
E V Sabaneeva
ABSTRACT
The technique of silver staining of the active nucleolar organizer regions is discussed. The specificity of the technique, the mechanism of the reaction, the nature and possible functions of argyrophilic protein(s) are considered. Special attention is paid to the fact, ascertained elsewhere, that the silver deposits in the nucleolus are related to the structure of "activated" (potentially active) ribosomal chromatin, rather than to the process of transcription itself. In this concern the supposition that the argyrophilic protein could interact with rDNA maintaining its non-nucleosomal conformation is under discussion. The possibility that the argyrophilic protein could be a component of the residual nucleolus is also taken into account. Whatever the function of the argyrophilic protein may be, this protein can be considered as a reliable marker for the active and "activated" ribosomal genes.
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O