Merck
Importance of magnesium chloride repletion after myocardial infarction.
The American journal of cardiology (1989-04-18)
J Sheehan
ABSTRACT
Data pertinent to the role of magnesium deficits in coronary artery disease and myocardial infarction are reviewed. Results of clinical, laboratory and epidemiologic studies indicate an association between magnesium deficiency and a poor prognostic outcome in patients who have had myocardial infarction. It therefore appears to be a reasonable prophylactic measure to monitor closely magnesium status in patients with coronary heart disease and other patients at risk of acute myocardial infarction, and to supplement with magnesium chloride when clinically necessary. In addition, recent studies provide supportive evidence that supplementation of magnesium chloride may reduce the incidence of fatal and nonfatal arrhythmias after an infarct.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O