Merck
  • The Jessner's-trichloroacetic acid peel. An enhanced medium-depth chemical peel.
The Jessner's-trichloroacetic acid peel. An enhanced medium-depth chemical peel.
Dermatologic clinics (1995-04-01)
G D Monheit
ABSTRACT
The Jessner's-trichloroacetic acid (TCA) peel is a procedure developed by Monheit to produce a safe, effective medium-depth chemical peel for the treatment of photoaged skin, actinic keratoses, and superficial acne scars. The technique of pretreatment degreasing and combination chemicals allow a deeper penetration for the 35% TCA, increasing its efficacy while preserving its safety. Technique and methods are reviewed.
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%