Merck
  • Role of beef and beef tallow, an enriched source of stearic acid, in a cholesterol-lowering diet.
Role of beef and beef tallow, an enriched source of stearic acid, in a cholesterol-lowering diet.
The American journal of clinical nutrition (1994-12-01)
M A Denke
ABSTRACT
The effects of stearic acid on metabolism must be evaluated for stearic acid as an isolated dietary constituent and for stearic acid as a component of a natural fat. Beef products are the most common source of dietary stearic acid in the United States. Two components of beef products, beef protein and beef fat, can potentially impart cholesterol-raising properties. Protein has minimal effects on cholesterol concentrations in humans, but studies suggest that beef fat raises serum cholesterol concentrations. Because beef fat is 19% stearic acid, the cholesterol-raising potential of beef is not as great as predicted by its total saturated fatty acid content. However, beef tallow is hypercholesterolemic compared with fats containing less cholesterol-raising saturated fatty acid. Therefore, curtailment of beef tallow in a cholesterol-lowering diet seems appropriate. Data suggest that lean beef is no more hypercholesterolemic than chicken or fish and, therefore, lean beef need not be eliminated from cholesterol-lowering diets.
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Stearic acid, European Pharmacopoeia (EP) Reference Standard
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, analytical standard