Merck
Clinical experiences with nimodipine in cerebral ischemia.
Journal of neural transmission. Supplementum (1994-01-01)
A Kakarieka, E H Schakel, J Fritze
ABSTRACT
Increased intracellular calcium is involved in the death of neurons associated with cerebral ischemia. After extensive experimental work, the calcium antagonist nimodipine has been investigated in several clinical trials in patients with disorders of the cerebral circulation. A short overview on trials in subarachnoid hemorrhage, head injury, ischemic stroke and cerebral resuscitation is given in this paper.
Sigma-Aldrich
Nimodipine
Nimodipine, European Pharmacopoeia (EP) Reference Standard
Supelco
Nimodipine, Pharmaceutical Secondary Standard; Certified Reference Material