Merck
  • The role of regenerative cell proliferation in chloroform-induced cancer.
The role of regenerative cell proliferation in chloroform-induced cancer.
Toxicology letters (1995-12-01)
B E Butterworth, M V Templin, S J Borghoff, R B Conolly, G L Kedderis, D C Wolf
ABSTRACT
Chloroform produces cancer by a nongenotoxic-cytotoxic mode of action, with no increased cancer risk expected at noncytotoxic doses. The default risk assessment for inhaled chloroform relies on liver tumor incidence from a gavage study with female B6C3F1 mice and estimates a virtually safe dose (VSD) at an airborne concentration of 0.000008 ppm of chloroform. In contrast, a 1000-fold safety factor applied to the NOAEL for liver cytotoxicity from inhalation studies yields a VSD of 0.01 ppm. This estimate relies on inhalation data and is more consistent with the mode of action of chloroform.
Product Description
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer