Merck
Pathogenesis of L-tryptophan eosinophilia myalgia syndrome.
Advances in experimental medicine and biology (1996-01-01)
E M Sternberg
ABSTRACT
Taken together, these studies suggest that many different etiologic agents alone or together may initiate the common final pathways of tissue pathologic response resulting in the clinical syndrome of eosinophilia, myalgias and fasciitis. Tryptophan itself may contribute to some of the scarring features of the illness, while impure L-tryptophan, and one or more of the impurities cause the characteristic features of the illness. The altered tryptophan metabolism in EMS is secondary to inflammation.
Product Description
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)