Merck
[Atenolol in treatment of patients with airway obstruction].
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego (1996-12-01)
R Chazan
ABSTRACT
It is well known that beta-2-blockers are contraindicated for patients with bronchial hyperactivity. Author discusses the clinical observations in relation to treatment with atenolol in circulatory system pathology.
Product Description
Supelco
Atenolol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
USP
Atenolol, United States Pharmacopeia (USP) Reference Standard
Atenolol, European Pharmacopoeia (EP) Reference Standard