Merck
Photoaging and tretinoin.
Dermatologic clinics (1998-05-20)
S Kang
ABSTRACT
Premature skin aging caused by repeated exposure to solar radiation is called photoaging. Although once considered an irreversible process, it is now established that photoaging can be treated by topical tretinoin. Both from carefully designed controlled clinical studies; and basic investigations into the mechanism by which tretinoin improves photoaged skin, our understanding of photoaging has been enhanced. This article highlights some of these studies which have contributed to our knowledge.
Product Description
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Tretinoin, European Pharmacopoeia (EP) Reference Standard
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material