Merck
Amine-chelated aryllithium reagents--structure and dynamics.
Journal of the American Chemical Society (2001-08-17)
H J Reich, W S Goldenberg, B O Gudmundsson, A W Sanders, K J Kulicke, K Simon, I A Guzei
ABSTRACT
Multinuclear NMR studies of five-membered-ring amine chelated aryllithium reagents 2-lithio-N,N-dimethylbenzylamine (1), the diethylamine and diisopropylamino analogues (2, 3), and the o-methoxy analogue (4), isotopically enriched in (6)Li and (15)N, have provided a detailed picture of the solution structures in ethereal solvents (usually in mixtures of THF and dimethyl ether, ether, and 2,5-dimethyltetrahydrofuran). The effect of cosolvents such as TMEDA, PMDTA, and HMPA has also been determined. All compounds are strongly chelated, and the chelation is not disrupted by these cosolvents. Reagents 1, 2, and 3 are dimeric in solvents containing a large fraction of THF. Below -120 degrees C, three chelation isomers of the dimers are detectable by NMR spectroscopy: one (A) with both nitrogens coordinated to one lithium of the dimer, and two (B and C) in which each lithium bears one chelating group. Dynamic NMR studies have provided rates and activation energies for the interconversion of the 1-A, 1-B, and 1-C isomers. They interconvert either by simple ring rotation, which interconverts B and C, or by amine decoordination (probably associative, DeltaG(++)(-93) = 8.5 kcal/mol), which can interconvert all of the isomers. The dimers of 1 are thermodynamically more stable than those of model systems such as phenyllithium, o-tolyllithium, or 2-isoamylphenyllithium (5, DeltaDeltaG > or = 3.3 kcal/mol). They are not detectably deaggregated by TMEDA or PMDTA, although HMPA causes partial deaggregation. The dimers are also more robust kinetically with rates of interaggregate exchange, measured by DNMR line shape analysis of the C-Li signal, orders of magnitude smaller than those of models (DeltaDeltaG(++) > or = 4.4 kcal/mol). Similarly, the mixed dimer of 1 and phenyllithium, 13, is kinetically more stable than the phenyllithium dimer by >2.2 kcal/mol. X-ray crystal structures of the TMEDA solvate of 1-A and the THF solvate of 3-B showed them to be dimeric and chelated in the solid state as well. Compound 4, which has a methoxy group ortho to the C-Li group, differs from the others in being only partially dimeric in THF, presumably for steric reasons. This compound is fully deaggregated by 1 equiv of HMPA. Excess HMPA leads to the formation of ca. 15% of a triple ion (4-T) in which both nitrogens appear to be chelated to the central lithium.