Merck
  • Simple diastereoselectivity of the BF3.OEt2-catalyzed vinylogous Mukaiyama aldol reaction of 2-(trimethylsiloxy)furans with aldehydes.
Simple diastereoselectivity of the BF3.OEt2-catalyzed vinylogous Mukaiyama aldol reaction of 2-(trimethylsiloxy)furans with aldehydes.
The Journal of organic chemistry (2005-04-23)
Carlos Silva López, Rosana Alvarez, Belén Vaz, Olalla Nieto Faza, Angel R de Lera
ABSTRACT
[reaction: see text] A comprehensive scan of the transition state space for the reaction of 2-(trimethylsiloxy)furan and methacrolein (24 combinations) offered a satisfactory explanation of the high like diastereoselectivity obtained experimentally in the Mukaiyama vinylogous aldol reaction of these and related partners. It was determined that the syn-gamma-hydroxyalkylbutenolides are formed preferentially following a g(+) orientation of the two reactants with the aldehyde in the s-trans conformation. Diastereoselectivity is shown to be caused by a combination of subtle effects favoring the formation of the like product.
Sigma-Aldrich
2-(Trimethylsiloxy)furan, 97%
Pricing and availability is not currently available.