Merck
  • CHIP suppresses polyglutamine aggregation and toxicity in vitro and in vivo.
CHIP suppresses polyglutamine aggregation and toxicity in vitro and in vivo.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2005-10-07)
Victor M Miller, Rick F Nelson, Cynthia M Gouvion, Aislinn Williams, Edgardo Rodriguez-Lebron, Scott Q Harper, Beverly L Davidson, Michael R Rebagliati, Henry L Paulson
ABSTRACT
Huntington's disease (HD) and other polyglutamine (polyQ) neurodegenerative diseases are characterized by neuronal accumulation of the disease protein, suggesting that the cellular ability to handle abnormal proteins is compromised. As both a cochaperone and ubiquitin ligase, the C-terminal Hsp70 (heat shock protein 70)-interacting protein (CHIP) links the two major arms of protein quality control, molecular chaperones, and the ubiquitin-proteasome system. Here, we demonstrate that CHIP suppresses polyQ aggregation and toxicity in transfected cell lines, primary neurons, and a novel zebrafish model of disease. Suppression by CHIP requires its cochaperone function, suggesting that CHIP acts to facilitate the solubility of mutant polyQ proteins through its interactions with chaperones. Conversely, HD transgenic mice that are haploinsufficient for CHIP display a markedly accelerated disease phenotype. We conclude that CHIP is a critical mediator of the neuronal response to misfolded polyQ protein and represents a potential therapeutic target in this important class of neurodegenerative diseases.
Sigma-Aldrich
Lactacystin, Synthetic, Lactacystin, Synthetic, CAS 1258004-00-0, is an irreversible inhibitor of 20S proteasome inhibitor (IC50 = 500 nM). A covalent inhibitor of the chymotrypsin & trypsin-like activities of proteasome.