Merck
  • Molecular cloning of human macrophage capping protein cDNA. A unique member of the gelsolin/villin family expressed primarily in macrophages.
Molecular cloning of human macrophage capping protein cDNA. A unique member of the gelsolin/villin family expressed primarily in macrophages.
The Journal of biological chemistry (1992-08-15)
G A Dabiri, C L Young, J Rosenbloom, F S Southwick
ABSTRACT
Macrophage capping protein (MCP) is a Ca(2+)-sensitive protein which reversibly blocks the barbed ends of actin filaments but does not sever preformed actin filaments. The human cDNA for MCP has been cloned and sequenced. The derived amino acid sequence predicts a polypeptide of 38.4 kDa. Human MCP expressed in Escherichia coli using a pET12a vector was functionally identical to the native protein purified from rabbit alveolar macrophages with respect to Ca2+ sensitivity and ability to block monomer exchange at the barbed end of actin filaments. Sequence comparison with other actin-binding protein sequences indicates that MCP is a member of the gelsolin/villin family of barbed end blocking proteins. Unlike gelsolin, this protein has a limited tissue distribution being detected primarily in macrophages where it was abundant, representing 0.9-1% of the total cytoplasmic protein. Northern blot analysis of U937 and HL60 cells differentiated to macrophage-like cells demonstrated that MCP message increases to 2.6 and greater than 7 times initial levels, respectively. Human MCP displays a 93% amino acid sequence identity with two recently described mouse proteins, gCap39 and Mbh1. Its abundance in macrophages and the corresponding increases in mRNA levels upon promyelocyte and monocyte development into macrophages indicate that MCP may play an important role in macrophage function.