Merck
Nutrition: the future of melanoma prevention?
Journal of the American Academy of Dermatology (2014-03-25)
Lana X Tong, Lorraine C Young
ABSTRACT
Melanoma is one of the deadliest forms of skin cancer, having a high metastatic potential and afflicting all age groups. The need for successful preventative measures is particularly urgent as metastatic melanoma is largely incurable. The beneficial role of nutrition and other natural compounds in the prevention and treatment of melanoma has been clearly demonstrated in the past, and is an exciting source for potential therapies in the future. We sought to review updates in the current literature regarding new developments in the relationship between nutrition and melanoma risk and treatment. Articles in the public domain regarding the impact of diet, grape seed proanthocyanidins, selenium, vitamin D, vitamin E, epigallocatechin-3-gallate, resveratrol, rosmarinic acid, lycopene, and fig latex on melanoma were included. Grape seed proanthocyanidins, epigallocatechin-3-gallate, resveratrol, rosmarinic acid, lycopene, and fig latex have demonstrated clear anticancer effects toward melanoma. The roles of selenium, vitamin D, and vitamin E, however, have been more controversial. None. The role of natural compounds in the future of melanoma prevention and treatment is promising and one that is worthy of further exploration.
Product Description
Supelco
(+)-Catechin, analytical standard
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Epigallocatechin gallate, primary reference standard
Supelco
(−)-Epigallocatechin gallate, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Resveratrol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Supelco
(−)-Epicatechin, analytical standard
Epicatechin, primary reference standard
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Lycopene, Redivivo, 10% FS, ~10% in corn oil, ≥95.0% (sum of isomers)
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Supelco
Lycopene, analytical standard
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)