Merck
  • The current position of precuffed expanded polytetrafluoroethylene bypass grafts in peripheral vascular surgery.
The current position of precuffed expanded polytetrafluoroethylene bypass grafts in peripheral vascular surgery.
Journal of vascular surgery (2014-03-19)
Jasper van der Slegt, Stijn L Steunenberg, Jeroen M W Donker, Eelco J Veen, Gwan H Ho, Hans G W de Groot, Lijckle van der Laan
ABSTRACT
Long-term results of precuffed expanded polytetrafluoroethylene (ePTFE) grafts used for peripheral bypass surgery are lacking. The aim of this study was to obtain the long-term outcomes of precuffed ePTFE grafts compared with autologous saphenous vein (ASV) grafts used in patients with peripheral arterial disease (PAD). A single-institution retrospective study of precuffed ePTFE and ASV graft performances in patients with PAD was undertaken between January 2004 and December 2012. Five-year primary patency, secondary patency, and limb salvage rates were determined by Kaplan-Meier analyses. A total of 467 bypass grafts were included in this study (169 precuffed ePTFE grafts and 298 ASV grafts). Secondary patency rates of ePTFE vs ASV at 1 and 5 years, respectively, were as follows: for 134 supragenicular femoropopliteal bypasses, 60% and 27% vs 89% and 85% (P < .05); for 190 infragenicular femoropopliteal bypasses, 40% and 25% vs 86% and 79% (P < .05); and for 84 femorocrural bypasses, 30% and 14% vs 50% and 50% (P < .05). Five-year limb salvage rates of ePTFE vs ASV for supragenicular femoropopliteal bypasses were 82% vs 94% (P = .16); for infragenicular femoropopliteal bypasses, 41% vs 92% (P < .05); and for femorocrural bypasses, 43% vs 64% (P = .06). ASV bypasses are still the first-choice conduit in peripheral bypass surgery performed in patients with PAD. Precuffed ePTFE bypasses are acceptable alternatives in the absence of adequate autologous vein.
Sigma-Aldrich
Polytetrafluoroethylene preparation, 60 wt % dispersion in H2O
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size