Merck
  • Gallic acid interacts with α-synuclein to prevent the structural collapse necessary for its aggregation.
Gallic acid interacts with α-synuclein to prevent the structural collapse necessary for its aggregation.
Biochimica et biophysica acta (2014-04-29)
Yanqin Liu, John A Carver, Antonio N Calabrese, Tara L Pukala
ABSTRACT
The accumulation of protein aggregates containing amyloid fibrils, with α-synuclein being the main component, is a pathological hallmark of Parkinson's disease (PD). Molecules which prevent the formation of amyloid fibrils or disassociate the toxic aggregates are touted as promising strategies to prevent or treat PD. In the present study, in vitro Thioflavin T fluorescence assays and transmission electron microscopy imaging results showed that gallic acid (GA) potently inhibits the formation of amyloid fibrils by α-synuclein. Ion mobility-mass spectrometry demonstrated that GA stabilises the extended, native structure of α-synuclein, whilst NMR spectroscopy revealed that GA interacts with α-synuclein transiently.
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Thioflavine S, practical grade
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland