Merck
  • Cytogenetic analysis of oral mucosa cells, induced by chlorhexidine, essential oils in ethanolic solution and triclosan mouthwashes.
Cytogenetic analysis of oral mucosa cells, induced by chlorhexidine, essential oils in ethanolic solution and triclosan mouthwashes.
Environmental research (2014-04-29)
Irene Ros-Llor, Pia Lopez-Jornet
ABSTRACT
The aim of this study was to evaluate DNA damage and cytokinetic defects, proliferative potential and cell death caused by the frequent use of mouthrinses containing chlorhexidine, triclosan and essential oils in ethanolic solution, compared to a placebo mouthwash. This double-blind, prospective, randomized clinical trial included 80 Caucasian patients. Subjects were divided into four groups: Group I used a mouthrinse, Triclosan; Group II used physiological saline; Group III used chlorhexidine; Group IV a mouthrinse with essential oils in ethanolic solution. All subjects used the mouthrinses for two weeks (15 ml, twice a day, rinsing for 30s). Two cell samples per subject were collected, before and after mouthrinse use (on day 0 and day 15). Samples were processed as follows: cell collection from cheeks with a cytobrush; cell centrifuge; slide preparation, fixation and staining; and fluorescent microscope analysis. 2000 exfoliated cells were screened for nuclear abnormalities, particularly the presence of micronuclei by means of cytome assay. No significant differences between study times (before and after use of mouthwash) were identified for any of the variables studied (p>0.05). Differences between mouthrinse groups were also compared but no significant differences were found (p>0.05). This study did not observe any genotoxic effect resulting from mouthrinse use.
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Chlorhexidine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®