Merck
  • Spontaneous cyclization of polypeptides with a penultimate Asp, Asn or isoAsp at the N-terminus and implications for cleavage by aminopeptidase.
Spontaneous cyclization of polypeptides with a penultimate Asp, Asn or isoAsp at the N-terminus and implications for cleavage by aminopeptidase.
The FEBS journal (2014-05-09)
Brian Lyons, Ann H Kwan, Roger Truscott
ABSTRACT
A cyclic product that forms spontaneously from peptides that contain a penultimate Asp, Asn or isoAsp residue at the N-terminus has been characterized. This 2,5-diketopiperazine derivative forms under physiological conditions and is stable, showing little degradation even following heating at 60 °C. A mechanism for its formation from Asn and Asp peptides is proposed that involves a succinimide or isoaspartate intermediate. A diketopiperazine-modified peptide was also detected in human lens extracts. Since peptides that contain the diketopiperazine moiety are not readily hydrolysed by leucine aminopeptidase, it is hypothesized that proteins and peptides modified in this way in the body may not readily be digested by the normal proteolytic machinery of cells.
Sigma-Aldrich
L-Asparagine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Asparagine, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
SAFC
L-Aspartic acid
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)