Merck
  • Design, formulation and optimization of valsartan transdermal gel containing iso-eucalyptol as novel permeation enhancer: preclinical assessment of pharmacokinetics in Wistar albino rats.
Design, formulation and optimization of valsartan transdermal gel containing iso-eucalyptol as novel permeation enhancer: preclinical assessment of pharmacokinetics in Wistar albino rats.
Expert opinion on drug delivery (2014-05-17)
Abdul Ahad, Mohd Aqil, Kanchan Kohli, Yasmin Sultana, Mohd Mujeeb
ABSTRACT
The aim of this study was to develop and optimize a transdermal gel formulation of valsartan using Box-Behnken design and to evaluate it for pharmacokinetic study. The independent variables were Carbopol 940 (X1), PEG 400 (X2) and ethanol (X3) while valsartan flux (Y1), Tlag (Y2) and gel viscosity (Y3) were the dependent variables. Iso-eucalyptol was added in all gel formulations as permeation enhancer except for control gel. It was observed that the permeation rate of valsartan significantly increased in direct proportion to the ethanol concentration, but significantly decreased in direct proportion to polymer concentration. Lag time and viscosity decreased in reverse proportion to ethanol concentration. The optimized valsartan gel formulation (VGF-OPT) yielded flux of 143.27 ± 7.11 µg/cm(2)/h and 27.55 ± 2.51 µg/cm(2)/h across rat and human cadaver skin, respectively. In vivo pharmacokinetic study of VGF-OPT-transdermal therapeutic system containing iso-eucalyptol showed a significant increase in the bioavailability (2.52 times) compared with oral formulation of valsartan by virtue of better permeation through Wistar rat skin. It was concluded that the developed transdermal gel accentuates the flux of valsartan and could be used as an antihypertensive dosage form for effective transdermal delivery of valsartan.
Product Description
Supelco
Valsartan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Valsartan, ≥98% (HPLC)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Eucalyptol, 99%
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Valsartan, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
Eucalyptol, analytical standard
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Cineole, European Pharmacopoeia (EP) Reference Standard