Merck
  • Melatonin and dopamine as biomarkers to optimize treatment in phenylketonuria: effects of tryptophan and tyrosine supplementation.
Melatonin and dopamine as biomarkers to optimize treatment in phenylketonuria: effects of tryptophan and tyrosine supplementation.
The Journal of pediatrics (2014-05-27)
Shoji Yano, Kathryn Moseley, Colleen Azen
ABSTRACT
To determine whether additional supplementation of tryptophan (Trp) and tyrosine (Tyr) improve serotonin and dopamine metabolism in individuals with phenylketonuria treated with large neutral amino acid (LNAA) tablets. Ten adult individuals with phenylketonuria participated in a randomized, double-blind, placebo-controlled cross-over study consisting of three 3-week phases: washout, treatment with LNAA tablets plus supplementation with either Trp and Tyr tablets or placebo, and LNAA tablets plus the alternate supplementation. An overnight protocol to measure blood melatonin, a serotonin metabolite in the pinealocytes, and urine 6-sulfatoxymelatonin and dopamine in first-void urine specimens was conducted after each phase. Serum melatonin and urine 6-sulfatoxymelatonin and dopamine levels were increased in the LNAA phase (LNAA plus placebo) compared with the washout phase. Serum melatonin and urine 6-sulfatoxymelatonin were not increased in the active phase (LNAA plus Trp + Tyr) compared with the LNAA phase, although plasma Trp:LNAA was increased compared with the LNAA phase. Among 7 subjects with a plasma Trp/LNAA >0.03, a negative correlation between urine 6-sulfatoxymelatonin and plasma phenylalanine levels was observed (r = -0.072). Urine dopamine levels and plasma Tyr:LNAA were increased in the active phase compared with the LNAA phase. Melatonin levels were not increased with the higher dose of Trp supplementation, but dopamine levels were increased with the higher dose of Tyr supplementation. Serotonin synthesis appears to be suppressed by high phenylalanine levels at the Trp hydroxylase level.
Product Description
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Supelco
Dopamine hydrochloride solution, 1.0 mg/mL in methanol with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®
USP
Melatonin, United States Pharmacopeia (USP) Reference Standard
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, FG
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Dopamine hydrochloride
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
USP
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard
Tryptophan, European Pharmacopoeia (EP) Reference Standard