Merck
  • Cadmium-induced oxidative stress tolerance in cadmium resistant Aspergillus foetidus: its possible role in cadmium bioremediation.
Cadmium-induced oxidative stress tolerance in cadmium resistant Aspergillus foetidus: its possible role in cadmium bioremediation.
Ecotoxicology and environmental safety (2014-05-20)
Shatarupa Chakraborty, Abhishek Mukherjee, Anisur Rahman Khuda-Bukhsh, Tapan Kumar Das
ABSTRACT
Toxic effects of cadmium (Cd) were examined on a cadmium-resistant strain of Aspergillus foetidus isolated from wastewater. The Cd removal potential was analyzed. The results indicated that the strain could tolerate up to 25 mM and 63 mM Cd in liquid and solid Czapek-Dox media, respectively. It efficiently removed Cd from liquid growth media and industrial wastewater by mycelial biosorption. The strain produced oxalic acid for the purpose of Cd bioleaching as confirmed by the presence of cadmium oxalate crystals on the mycelial surface. Intracellular proline contents and the antioxidative enzyme activities increased up to a certain level to detoxify the overproduced free radicals. These data indicate that the strain has inherent mechanisms to grow in Cd contaminated environment, tolerate high Cd doses and high Cd uptake potential which are pre-requisite for acting as a suitable candidate for Cd bioremediation.
Product Description
Cadmium, foil, light tested, 25x25mm, thickness 0.025mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.001mm, 99.7+%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, wire reel, 0.1m, diameter 2.0mm, hard, 99.99+%
Cadmium, wire reel, 0.1m, diameter 0.70mm, as drawn, 99.99+%
Cadmium, foil, 50mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.7+%
Cadmium, tube, 1000mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.025mm, 99.7+%
Cadmium, tube, 100mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, light tested, 100x100mm, thickness 0.025mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.005mm, 99.7+%
Cadmium, rod, 150mm, diameter 10.0mm, as drawn, 99.99+%
Cadmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, not light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, wire reel, 0.2m, diameter 2.0mm, hard, 99.99+%
Cadmium, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 6mm disks, thickness 0.005mm, 99.7+%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.002mm, 99.7+%