Merck
  • Targeting the disordered C terminus of PTP1B with an allosteric inhibitor.
Targeting the disordered C terminus of PTP1B with an allosteric inhibitor.
Nature chemical biology (2014-05-23)
Navasona Krishnan, Dorothy Koveal, Daniel H Miller, Bin Xue, Sai Dipikaa Akshinthala, Jaka Kragelj, Malene Ringkjøbing Jensen, Carla-Maria Gauss, Rebecca Page, Martin Blackledge, Senthil K Muthuswamy, Wolfgang Peti, Nicholas K Tonks
ABSTRACT
PTP1B, a validated therapeutic target for diabetes and obesity, has a critical positive role in HER2 signaling in breast tumorigenesis. Efforts to develop therapeutic inhibitors of PTP1B have been frustrated by the chemical properties of the active site. We define a new mechanism of allosteric inhibition that targets the C-terminal, noncatalytic segment of PTP1B. We present what is to our knowledge the first ensemble structure of PTP1B containing this intrinsically disordered segment, within which we identified a binding site for the small-molecule inhibitor MSI-1436. We demonstrate binding to a second site close to the catalytic domain, with cooperative effects between the two sites locking PTP1B in an inactive state. MSI-1436 antagonized HER2 signaling, inhibited tumorigenesis in xenografts and abrogated metastasis in the NDL2 mouse model of breast cancer, validating inhibition of PTP1B as a therapeutic strategy in breast cancer. This new approach to inhibition of PTP1B emphasizes the potential of disordered segments of proteins as specific binding sites for therapeutic small molecules.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Roche
X-tremeGENE HP DNA Transfection Reagent, High-performance polymer reagent for transfecting many cell lines
Supelco
Spermine, analytical standard
Sigma-Aldrich
Spermine, ≥99.0% (GC)
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
Spermine, suitable for cell culture, BioReagent