Merck
  • A cytosolic glutathione s-transferase, GST-theta from freshwater prawn Macrobrachium rosenbergii: molecular and biochemical properties.
A cytosolic glutathione s-transferase, GST-theta from freshwater prawn Macrobrachium rosenbergii: molecular and biochemical properties.
Gene (2014-06-01)
Jesu Arockiaraj, Annie J Gnanam, Rajesh Palanisamy, Prasanth Bhatt, Venkatesh Kumaresan, Mukesh Kumar Chaurasia, Mukesh Pasupuleti, Harikrishnan Ramaswamy, Abirami Arasu, Akila Sathyamoorthi
ABSTRACT
Glutathione S-transferases play an important role in cellular detoxification and may have evolved to protect cells against reactive oxygen metabolites. In this study, we report the molecular characterization of glutathione s-transferase-theta (GST-θ) from freshwater prawn Macrobrachium rosenbergii. A full length cDNA of GSTT (1417 base pairs) was isolated and characterized bioinformatically. Exposure to virus (white spot syndrome baculovirus or M. rosenbergii nodovirus), bacteria (Aeromonas hydrophila or Vibrio harveyi) or heavy metals (cadmium or lead) significantly increased the expression of GSTT (P<0.05) in hepatopancreas. Recombinant GST-θ with monochlorobimane substrate had an optimum activity at pH7.5 and 35 °C. Furthermore recombinant GST-θ activity was abolished by the denaturants triton X-100, Gua-HCl, Gua-thiocyanate, SDS and urea in a dose-dependent manner. Overall, the results suggest a potential role for M. rosenbergii GST-θ in detoxification and possibly conferring immune protection.
Product Description
Lead, foil, 2m coil, thickness 0.125mm, as rolled, 99.95%
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.02mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.008mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.009mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.04mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.02mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.04mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.05mm, 99.99+%
Lead, foil, not light tested, 300x300mm, thickness 0.016mm, as rolled, 96% (Sn 1.5%, Sb 2.5%)
Lead, wire reel, 1m, diameter 0.75mm, as drawn, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.02mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.02mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.009mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.04mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.007mm, 99.99+%
Lead, foil, not light tested, 100x100mm, thickness 0.01mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.008mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.025mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.004mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.05mm, as rolled, 96% (Sn 1.5%, Sb 2.5%)
Lead, tube, 200mm, outside diameter 6.78mm, inside diameter 4.7mm, wall thickness 1.04mm, 99.9%
Lead, foil, not light tested, 100x100mm, thickness 0.008mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.006mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 0.5m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.25mm, as rolled, 99.99+%