Merck
  • Photochemical performance of thylakoid membrane in lead-treated Nymphoides peltatum.
Photochemical performance of thylakoid membrane in lead-treated Nymphoides peltatum.
Bulletin of environmental contamination and toxicology (2014-06-16)
Xuqiang Qiao, Guoxin Shi, Zhenzhen Zheng, Min Huang, Haiyan Yang
ABSTRACT
Photochemical responses in the thylakoid membrane of Nymphoides peltatum to increasing lead (Pb) concentrations were investigated after 5 days of exposure. Pb accumulation increased in a concentration-dependent manner, with a maximum of 118.44 μg g(-1) at 100 μM. Nutrients (Zn, Mg, Mo, Ca, Fe and Mn), ATPase activity and pigment generally increased progressively at Pb concentrations of 12.5 and 25 μM, but then declined at concentrations of 50 and 100 μM. Moreover, Pb stress induced an increase in chlorophyll (Chl) a/b ratio in a different extent. No outstanding changes were observed in several Chl a fluorescence parameters at low Pb concentrations (12.5 and 25 μM), while significant changes (p < 0.05) were observed in these parameters at higher concentrations. The alterations of nutrients, ATPase activity and pigment content were associated with disturbances in the thylakoid membrane, indicated by the quenching of Chl a fluorescence. These results were indicative of a disarray in photochemical activities exerted by Pb phytotoxicity.
Sigma-Aldrich
Chlorophyll b from spinach, ≥90% (HPLC), ≤0.5% Chlorophyll a
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Lead, foil, not light tested, 50x50mm, thickness 0.004mm, 99.99+%
Lead, foil, 8mm disks, thickness 0.03mm, 99.99+%
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 8mm disks, thickness 0.006mm, 99.99+%
Lead, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 300x300mm, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 500x1000mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 5m coil, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 8mm disks, thickness 0.007mm, 99.99+%
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 8mm disks, thickness 0.05mm, 99.99+%
Lead, foil, 25x25mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 5m coil, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 1m coil, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 0.5m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 20m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Lead, foil, 10m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.025mm, 99.99+%