Merck
  • Characterization of the lipid binding properties of Otoferlin reveals specific interactions between PI(4,5)P2 and the C2C and C2F domains.
Characterization of the lipid binding properties of Otoferlin reveals specific interactions between PI(4,5)P2 and the C2C and C2F domains.
Biochemistry (2014-07-08)
Murugesh Padmanarayana, Nicole Hams, Lee C Speight, E James Petersson, Ryan A Mehl, Colin P Johnson
ABSTRACT
Otoferlin is a transmembrane protein consisting of six C2 domains, proposed to act as a calcium sensor for exocytosis. Although otoferlin is believed to bind calcium and lipids, the lipid specificity and identity of the calcium binding domains are controversial. Further, it is currently unclear whether the calcium binding affinity of otoferlin quantitatively matches the maximal intracellular presynaptic calcium concentrations of ∼30-50 μM known to elicit exocytosis. To characterize the calcium and lipid binding properties of otoferlin, we used isothermal titration calorimetry (ITC), liposome sedimentation assays, and fluorescence spectroscopy. Analysis of ITC data indicates that with the exception of the C2A domain, the C2 domains of otoferlin bind multiple calcium ions with moderate (Kd = 25-95 μM) and low affinities (Kd = 400-700 μM) in solution. However, in the presence of liposomes, the calcium sensitivity of the domains increased by up to 10-fold. It was also determined that calcium enhanced liposome binding for domains C2B-C2E, whereas the C2F domain bound liposomes in a calcium-independent manner. Mutations that abrogate calcium binding in C2F do not disrupt liposome binding, supporting the conclusion that the interaction of the C2F domain with phosphatidylserine is calcium-independent. Further, domains C2C and C2F, not domains C2A, C2B, C2D, and C2E, bound phosphatidylinositol 4,5-bisphosphate 1,2-dioleoyl-sn-glycero-3-phospho(1'-myoinositol-4',5'-bisphosphate) [PI(4,5)P2], which preferentially steered them toward liposomes harboring PI(4,5)P2. Remarkably, lysine mutations L478A and L480A in C2C selectively weaken the PI(4,5)P2 interaction while leaving phosphatidylserine binding unaffected. Finally, shifts in the emission spectra of an environmentally sensitive fluorescent unnatural amino acid indicate that the calcium binding loops of the C2F domain directly interact with the lipid bilayer of negatively charged liposomes in a calcium-independent manner. On the basis of these results, we propose that the C2F and C2C domains of otoferlin preferentially bind PI(4,5)P2 and that PI(4,5)P2 may serve to target otoferlin to the presynapse in a calcium-independent manner. This positioning would facilitate fast calcium-dependent exocytosis at the hair cell synapse.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, puriss. p.a., powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Carbon, foil, 50x50mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 300mm, diameter 10.0mm, graphite, 100%
Carbon, foil, 150x150mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%
Carbon, rod, 300mm, diameter 2.0mm, graphite, 100%
Carbon, rod, 100mm, diameter 25mm, graphite, 100%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Carbon, foil, not light tested, 300x300mm, thickness 0.125mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 50mm, diameter 50mm, graphite, 99.95%
Carbon, foil, 25x25mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%
Carbon, rod, 50mm, diameter 25mm, graphite, 100%
Carbon, rod, 150mm, diameter 25mm, graphite, 99.95%
Carbon, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Carbon, foil, 50mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, microleaf, 50x70mm, thinness 1.25μm, specific density 250μg/cm2, 99.997%
Carbon, foil, 500x500mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 0.1mm, pyrolytic graphite, 99.99%
Carbon, rod, 300mm, diameter 25mm, graphite, 99.95%