Merck
  • Valproic acid enhances the anti-tumor effect of pegylated interferon-α towards pancreatic cancer cell lines.
Valproic acid enhances the anti-tumor effect of pegylated interferon-α towards pancreatic cancer cell lines.
Anticancer research (2014-07-02)
Koji Sugimoto, Mitsuo Shimada, Toru Utsunomiya, Yuji Morine, Satoru Imura, Tetsuya Ikemoto, Shuichi Iwahashi
ABSTRACT
Valproic acid (VPA) acts as a specific inhibitor of class I HDACs and it use has been proven to be safe since a long time. In the present study, we investigated the effect of VPA in the combination with pegylated interferon-α (PEG-IFNα) in inhibition of cell proliferation of human pancreatic cancer cell lines. VPA enhanced the effect of PEG-IFNα, and the effect was decreased by the caspase inhibitor. VPA alone and VPA in combination with PEG-IFNα increased the expression of interferon-α receptor and interferon regulatory factor 8. The combination of VPA and PEG-IFNα can be useful for the treatment of pancreatic cancer.
Sigma-Aldrich
2-Propylpentanoic acid
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Valproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®