Merck
  • Loading of titanium implants with hydrophilic endosteal surface 3 weeks after insertion: clinical and radiological outcome of a 12-month prospective clinical trial.
Loading of titanium implants with hydrophilic endosteal surface 3 weeks after insertion: clinical and radiological outcome of a 12-month prospective clinical trial.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-07-20)
Robert M Hinkle, Stephen R Rimer, Michael H Morgan, Paul Zeman
ABSTRACT
The aim of this study was to test whether early loading (21 days after insertion) of implants with a hydrophilic surface is safe and reliable in a general clinical practice setting. This was a prospective multicenter clinical trial conducted in 3 independent surgical practices. Patients were selected according to predefined inclusion and exclusion criteria. Implants were placed in posterior regions into healed alveoli and native bone using a 1-stage (unsubmerged) surgical protocol. Loading was to be performed 21 days thereafter. Patients were followed clinically and radiographically for 1 year. Twenty-one patients who received 23 implants were followed for 1 year. None of the osseointegrated implants were lost. One broken provisional crown was reported during the trial period. The crestal bone remained remarkably stable (ie, approximately 1 mm below the microgap). The most coronal bone-to-implant contact stabilized at the level of the first implant thread just beneath the machined collar. Within the limitations of this clinical study, the authors confirmed that loading of implants with a hydrophilic surface 3 weeks after placement appears to be a safe and predictable treatment option. No crestal bone loss was observed 1 year after implant placement. Therefore, it appears that crestal bone loss is dependent on the design features of the specific implant line.
Product Description
Titanium, IRMM®, certified reference material, 0.1 mm foil
Titanium, rod, 100mm, diameter 8.0mm, as drawn, 99.99+%
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Titanium, rod, 100mm, diameter 20mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Titanium, tube, 1000mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 20mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Titanium, sphere, 200pcs, diameter 3mm, 99.6+%
Titanium, tube, 1000mm, outside diameter 3.0mm, inside diameter 1.5mm, wall thickness 0.75mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 6mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, rod, 50mm, diameter 9.5mm, annealed, 99.99+%
Titanium, rod, 25mm, diameter 9.5mm, annealed, 99.99+%
Titanium, tube, 1000mm, outside diameter 6.1mm, inside diameter 5.1mm, wall thickness 0.5mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 16mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 50mm, diameter 16mm, as drawn, 99.99+%
Titanium, rod, 25mm, diameter 20mm, as drawn, 99.99+%
Titanium, tube, 200mm, outside diameter 3.0mm, inside diameter 1.5mm, wall thickness 0.75mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 10.3mm, inside diameter 8.7mm, wall thickness 0.8mm, annealed, 99.6+%
Titanium, wire reel, 2m, diameter 0.5mm, as drawn, 99.99+%
Titanium, wire reel, 1m, diameter 1.0mm, as drawn, 99.6+%
Titanium, wire reel, 500m, diameter 0.25mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 1.5mm, as drawn, 99.6+%