Merck
  • Ultrasound-assisted surfactant-enhanced emulsification microextraction for the determination of Cd and Ni in tea and water samples.
Ultrasound-assisted surfactant-enhanced emulsification microextraction for the determination of Cd and Ni in tea and water samples.
Environmental technology (2014-08-26)
Maryam Ezoddin, Tayebeh Taghizadeh, Behrooz Majidi
ABSTRACT
A microextraction method based on ultrasound-assisted surfactant-enhanced emulsification using solidification of a floating organic droplet (UASEME-SFO) was evaluated for simultaneous determination of Cd and Ni in water and tea samples followed by flame atomic absorption spectrometry. In the UASEME-SFO technique, Triton X-100 was used as an emulsifier to accelerate the emulsification of the extraction solvent into a sample solution and hasten the mass transfer of the analytes. Analytes form a complex and are extracted into 1-dodecanol which was used as an extraction solvent. Some parameters such as type and volume of the extraction solvent, the type and concentration of the surfactant, ultrasound extraction time, reagent concentration, centrifuge conditions and salt concentration were investigated. Under optimum conditions, calibration curves were linear in the range of 0.3-100 and 0.6-180 microg L(-1) with detection limits of 0.11 and 0.20 microg L(-1) for Cd and Ni, respectively. The accuracy of the method was confirmed by parallel analyses using the certified reference material of water and tea samples. The recoveries of the analytes in tea leaves, tea infusions and water samples were in the range of 96.5-105.1%.
Product Description
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Nickel, foil, 10mm disks, thickness 0.1mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.20mm, annealed, 99%
Nickel, foil, 10mm disks, thickness 0.5mm, as rolled, 100%
Nickel, rod, 200mm, diameter 30mm, 99%
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Nickel, rod, 500mm, diameter 20.0mm, 99%
Nickel, foil, 50mm disks, thickness 0.01mm, 99.95%
Nickel, foil, 50mm disks, thickness 0.025mm, 99.9%
Nickel, rod, 200mm, diameter 40mm, 99%
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Nickel, rod, 100mm, diameter 20.0mm, 99%
Nickel, rod, 200mm, diameter 12.7mm, 99.98%
Nickel, rod, 200mm, diameter 1.5mm, 99.99+%
Nickel, foil, 10mm disks, thickness 0.05mm, as rolled, 100%
Nickel, rod, 200mm, diameter 6.35mm, 99%
Nickel, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Nickel, tube, 1000mm, outside diameter 1.8mm, inside diameter 1.6mm, wall thickness 0.1mm, hard, 99.5%
Nickel, rod, 100mm, diameter 16mm, 99.99+%
Nickel, tube, 1000mm, outside diameter 4.5mm, inside diameter 3.154mm, wall thickness 0.673mm, hard, 99.5%
Nickel, tube, 1000mm, outside diameter 0.8mm, inside diameter 0.3mm, wall thickness 0.25mm, as drawn, 99.5%
Nickel, foil, 10mm disks, thickness 0.30mm, as rolled, 99.98%
Nickel, foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%
Nickel, rod, 200mm, diameter 20.0mm, 99%
Nickel, foil, 10mm disks, thickness 0.125mm, as rolled, 100%
Nickel, foil, 15mm disks, thickness 0.0125mm, annealed, 99%
Nickel, tube, 1000mm, outside diameter 0.8mm, inside diameter 0.64mm, wall thickness 0.08mm, as drawn, 99.5%