Merck
  • Reproductive performance and oviductal expression of avidin and avidin-related protein-2 in young and old broiler breeder hens orally exposed to supplementary biotin.
Reproductive performance and oviductal expression of avidin and avidin-related protein-2 in young and old broiler breeder hens orally exposed to supplementary biotin.
Poultry science (2014-07-09)
H Daryabari, A Akhlaghi, M J Zamiri, G Rahimi Mianji, Z Ansari Pirsaraei, H Deldar, A N Eghbalian
ABSTRACT
Published data on the probable involvement of avidin and avidin-related protein-2 (AVR2) in sustaining sperm viability in sperm storage tubules in 38-wk-old turkeys, and the high affinity of avidin or its analogs to biotin suggest that supplementary biotin may increase oviductal avidin and AVR2 expression, thereby attenuating the adverse effect of aging on hen reproductive performance. Broiler breeder hens (n = 120) were randomly assigned to receive 0 (T0), 0.30 (T1), or 0.45 (T2) mg of biotin/L of drinking water from 30 to 33 (young) and 53 to 56 (old) wk of age, and artificially inseminated to determine their reproductive performance. At the end of each period of biotin administration, 8 hens from each treatment group were killed for RNA extraction from the uterovaginal junction. Egg production was lower in the old hens (44%) compared with the young ones (82%), and biotin supplementation increased egg production only in the latter. Administering supplementary biotin to young hens increased their oviductal expression of AVR2, which was much higher in the old hens (1.0 and 4.6 for young and old groups, respectively). Fertility rate was not different between young and old hens, and was increased (4.4%) at the higher level of biotin supplementation. Hatchability and hatchling quality were not affected by biotin supplementation. Embryonic mortality between 17 to 21 d of incubation was higher in young (5.2%) compared with old (1.4%) birds. Egg fertility rate showed a moderate correlation (P < 0.05) with avidin (r = 0.59) and AVR2 (r = 0.55) expression in the young-age group, and very low correlations in old-age group (0.04 and 0.17). Regardless of the hen's age, the correlation coefficient of hatchability with avidin or AVR2 expression was very low (-0.16 and 0.18). Overall, the effect of biotin supplementation on AVR2 expression, and the relationship between biotin administration and oviductal expression of avidin and AVR2 was dependent on the hen's age, being higher in the young hens.
Sigma-Aldrich
ExtrAvidin®−FITC, buffered aqueous solution
Sigma-Aldrich
ExtrAvidin®−Cy3, buffered aqueous solution
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
ExtrAvidin® –Alkaline Phosphatase, buffered aqueous solution