Merck
  • Uptake rate behavior of tube-type passive samplers for volatile organic compounds under controlled atmospheric conditions
Uptake rate behavior of tube-type passive samplers for volatile organic compounds under controlled atmospheric conditions
Atmospheric Environment (2011)
Walgraeve, C., et al.