Merck
  • Ligand-induced trafficking of the sphingosine-1-phosphate receptor EDG-1.
Ligand-induced trafficking of the sphingosine-1-phosphate receptor EDG-1.
Molecular biology of the cell (1999-04-10)
C H Liu, S Thangada, M J Lee, J R Van Brocklyn, S Spiegel, T Hla
ABSTRACT
The endothelial-derived G-protein-coupled receptor EDG-1 is a high-affinity receptor for the bioactive lipid mediator sphingosine-1-phosphate (SPP). In the present study, we constructed the EDG-1-green fluorescent protein (GFP) chimera to examine the dynamics and subcellular localization of SPP-EDG-1 interaction. SPP binds to EDG-1-GFP and transduces intracellular signals in a manner indistinguishable from that seen with the wild-type receptor. Human embryonic kidney 293 cells stably transfected with the EDG-1-GFP cDNA expressed the receptor primarily on the plasma membrane. Exogenous SPP treatment, in a dose-dependent manner, induced receptor translocation to perinuclear vesicles with a tau1/2 of approximately 15 min. The EDG-1-GFP-containing vesicles are distinct from mitochondria but colocalize in part with endocytic vesicles and lysosomes. Neither the low-affinity agonist lysophosphatidic acid nor other sphingolipids, ceramide, ceramide-1-phosphate, or sphingosylphosphorylcholine, influenced receptor trafficking. Receptor internalization was completely inhibited by truncation of the C terminus. After SPP washout, EDG-1-GFP recycles back to the plasma membrane with a tau1/2 of approximately 30 min. We conclude that the high-affinity ligand SPP specifically induces the reversible trafficking of EDG-1 via the endosomal pathway and that the C-terminal intracellular domain of the receptor is critical for this process.