Merck
Sensitivity of pressure sensors enhanced by doping silver nanowires.
Sensors (Basel, Switzerland) (2014-06-06)
Baozhang Li, Chengyi Xu, Jianming Zheng, Chunye Xu
ABSTRACT
We have developed a highly sensitive flexible pressure sensor based on a piezopolymer and silver nanowires (AgNWs) composite. The composite nanofiber webs are made by electrospinning mixed solutions of poly(inylidene fluoride) (PVDF) and Ag NWs in a cosolvent mixture of dimethyl formamide and acetone. The diameter of the fibers ranges from 200 nm to 500 nm, as demonstrated by SEM images. FTIR and XRD results reveal that doping Ag NWs into PVDF greatly enhances the content of β phase in PVDF. This β phase increase can be attributed to interactions between the Ag NWs and the PVDF matrix, which forces the polymer chains to be embedded into the β phase crystalline. The sensitivity of the pressure sensors agrees well with the FTIR and XRD characteristics. In our experiments, the measured sensitivity reached up to 30 pC/N for the nanofiber webs containing 1.5 wt% Ag NWs, which is close to that of poly(vinylidene fluoride-trifluoroethylene) [P(VDF-TrFE), (77/23)]. This study may provide a new method of fabricating high performance flexible sensors at relatively low cost compared with sensors based on [P(VDF-TrFE), (77/23)].
Sigma-Aldrich
Poly(vinylidene fluoride), powder
Sigma-Aldrich
Ethylene glycol 5 M solution
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Supelco
Acetone, analytical standard
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%