Merck
  • Dichloromethane activation. Direct methylenation of ketones and aldehydes with CH2Cl2 promoted by Mg/TiCl4/THF.
Dichloromethane activation. Direct methylenation of ketones and aldehydes with CH2Cl2 promoted by Mg/TiCl4/THF.
Organic letters (2004-12-21)
Tu-Hsin Yan, Chia-Chung Tsai, Ching-Ting Chien, Chia-Ching Cho, Pei-Chen Huang
ABSTRACT
[reaction: see text] This Mg-TiCl4-promoted CH2-transfer reaction of CH2Cl2 represents an extremely simple, practical, and efficient methylenation of a variety of ketones and aldehydes, especially in enolizable or sterically hindered ketones such as 2,2-dimethylcyclohexanone, camphor, and fenchone.
Sigma-Aldrich
2,2-Dimethylcyclohexanone, 92%
Pricing and availability is not currently available.