Merck
  • Palmitate induces mRNA translation and increases ER protein load in islet β-cells via activation of the mammalian target of rapamycin pathway.
Palmitate induces mRNA translation and increases ER protein load in islet β-cells via activation of the mammalian target of rapamycin pathway.
Diabetes (2014-05-20)
Masayuki Hatanaka, Bernhard Maier, Emily K Sims, Andrew T Templin, Rohit N Kulkarni, Carmella Evans-Molina, Raghavendra G Mirmira
ABSTRACT
Saturated free fatty acids (FFAs) have complex effects on the islet β-cell, acutely promoting adaptive hyperplasia but chronically impairing insulin release. The acute effects of FFAs remain incompletely defined. To elucidate these early molecular events, we incubated mouse β-cells and islets with palmitate and then studied mRNA translation by polyribosomal profiling and analyzed signaling pathways by immunoblot analysis. We found that palmitate acutely increases polyribosome occupancy of total RNA, consistent with an increase in mRNA translation. This effect on translation was attributable to activation of mammalian target of rapamycin (mTOR) pathways via L-type Ca(2+) channels but was independent of insulin signaling. Longer incubations led to depletion of polyribosome-associated RNA, consistent with activation of the unfolded protein response (UPR). Pharmacologic inhibition of mTOR suppressed both the acute effects of palmitate on mRNA translation and the chronic effects on the UPR. Islets from mice fed a high-fat diet for 7 days showed increases in polyribosome-associated RNA and phosphorylation of S6K, both consistent with activation of mTOR. Our results suggest that palmitate acutely activates mRNA translation and that this increase in protein load contributes to the later UPR.
Product Description
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, lyophilized powder, ≥350 units/mg protein, Protein ≥10 % by biuret
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, Type F-300, lyophilized powder, ≥130 units/mg protein (biuret)
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Palmitic acid, natural, 98%, FG