Merck
  • The influence of a glucosyltransferase, encoded by gtfP, on biofilm formation by Streptococcus sanguinis in a dual-species model.
The influence of a glucosyltransferase, encoded by gtfP, on biofilm formation by Streptococcus sanguinis in a dual-species model.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2014-03-19)
Yasuo Yoshida, Hiroyasu Konno, Keiji Nagano, Yuki Abiko, Yoshinori Nakamura, Yoshinobu Tanaka, Fuminobu Yoshimura
ABSTRACT
Streptococcus sanguinis is an early colonizer of tooth surfaces and forms biofilms with other species of microorganisms. In vitro, S. sanguinis produces water-soluble glucans from sucrose and releases them into the culture supernatant; however, the role played by these glucans in biofilm formation is unclear. The present study examined both the effect of glucans on biofilm formation by S. sanguinis and the proportion of this bacterial species within the biofilms. Inactivation of the gtfP gene, annotated as glucosyltransferase in the S. sanguinis genome database, caused a marked reduction in the amount of water-soluble glucans in the culture supernatant, but not in the amount of water-insoluble glucans expressed on the bacterial cell surface. Scanning electron microscopy revealed that wild-type S. sanguinis, but not the gtfP-deficient mutant, produced large amounts of sticky material in the presence of 1% (w/v) sucrose. In addition, biofilm production by wild-type bacteria was greater than that by the mutant strain. By contrast, co-culture of mutant bacteria with Streptococcus mutans, S. sobrinus, S. oralis, S. gordonii, S. anginosus, or S. salivarius showed that inactivating the gtfP gene had little effect on the amount of biofilm produced. Furthermore, inactivating the gtfP gene did not greatly alter the proportion of S. sanguinis in the biofilms formed by the co-cultures. Thus, despite the role of S. sanguinis glucosyltransferase in formation of water-soluble glucans and biofilms in monoculture, the functional gene contributed little to biofilms in co-culture experiments.
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sucrose, European Pharmacopoeia (EP) Reference Standard