Merck
  • FTIR spectroscopy offers hints towards widespread molecular changes in cobalt-acclimated freshwater bacteria.
FTIR spectroscopy offers hints towards widespread molecular changes in cobalt-acclimated freshwater bacteria.
Aquatic toxicology (Amsterdam, Netherlands) (2014-06-26)
Mehmet Kardas, Ayse Gul Gozen, Feride Severcan
ABSTRACT
High concentrations of heavy metals can be toxic for bacteria. However, after prolonged exposure, bacteria can become acclimated and begin to be able to grow in the presence of heavy metals. Acclimation can involve alterations of metabolism and molecular structures. Our aim was to examine these alterations in cobalt-acclimated bacteria via attenuated total reflection Fourier transform infrared (ATR-FTIR) spectroscopy on viable samples. Bacillus sp. and Pseudomonas sp. isolated from a temperate shallow lake and a well-established strain of E. coli were investigated. Our results revealed consistent, wide-spread changes in cell membrane and cell wall dynamics of Bacillus sp. and E. coli, including a decrease in peptidoglycan content of Bacillus sp. and increased lipid ordering of the membrane in both bacteria. Furthermore, a decrease in RNA and protein concentrations of Bacillus sp. was measured. All three bacteria studied showed a decrease in conformational freedom of proteins following cobalt acclimation. Interestingly, both Bacillus sp. and E. coli showed slight but significant alterations in their DNA conformations which might imply a methylation-mediated memory formation leading to epigenetic modulation for cobalt adaptation.
Product Description
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 50x50mm, thickness 0.025mm, 99.9%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Cobalt, wire reel, 2m, diameter 0.05mm, hard, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Cobalt, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Cobalt, foil, 10mm disks, thickness 0.009mm, 99.9%
Cobalt, foil, 15mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.025mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, 6mm disks, thickness 0.006mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 4mm disks, thickness 0.025mm, 99.9%
Cobalt, microfoil, disks, 10mm, thinness 0.25μm, specific density 222μg/cm2, permanent mylar 3.5μm support, 99.9%
Cobalt, foil, 6mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.9%