Merck
  • Effect of thiobencarb and pretilachlor on microorganisms in relation to mineralization of C and N in the Gangetic alluvial soil of West Bengal.
Effect of thiobencarb and pretilachlor on microorganisms in relation to mineralization of C and N in the Gangetic alluvial soil of West Bengal.
Environmental monitoring and assessment (2014-07-06)
Sourav Bhowmick, Ritwika Das, Amal Chandra Das
ABSTRACT
An experiment was conducted under laboratory conditions to investigate the effect of two pre-emergence herbicides, viz., thiobencarb (at 1.5 and 4.5 kg active ingredient (a.i.) ha(-1)) and pretilachlor (at 0.5 and 1.5 kg a.i. ha(-1)), on the growth and multiplication of some microorganisms (bacteria, actinomycetes and fungi) in relation to transformations and availability of C and N in the Gangetic alluvial soil (Typic Haplustept) of West Bengal, India. Application of both the herbicides, in general, significantly increased microbial biomass, resulting in greater retention, mineralization and availability of oxidizable organic C and N in soil, and the stimulations were more pronounced when the herbicides were applied at their lower concentrations (recommended field application rates), more so with thiobencarb, as compared to pretilachlor. Compared to untreated control soil, the application of thiobencarb at lower concentration increased the proliferation of total bacteria, actinomycetes and fungi by 57.3, 36.6 and 55.2%, respectively, and released the highest amount (40.2%) of soluble NO₃(-) in soil, while pretilachlor at field application rate induced the growth and multiplication of bacteria and fungi by 58.3 and 17.6%, respectively. Irrespective of the concentrations, the stimulations were at par for both the herbicides towards the retention of oxidizable organic C, total N and exchangeable NH₄(+) in soil.
Product Description
Supelco
Thiobencarb, PESTANAL®, analytical standard
Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon
Carbon, rod, 300mm, diameter 2.0mm, graphite, 100%
Carbon, foil, 100x100mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, foil, 25x25mm, thickness 0.5mm, pyrolytic graphite, 99.99%
Carbon, rod, 100mm, diameter 1.0mm, graphite, 99.95%
Carbon, rod, 50mm, diameter 0.5mm, graphite, 99.95%
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 100x100mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon