Merck
  • Immobilization of B, F, Cr, and As in alkaline coal fly ash through an aging process with water.
Immobilization of B, F, Cr, and As in alkaline coal fly ash through an aging process with water.
Environmental monitoring and assessment (2014-07-10)
Yasumasa Ogawa, Kento Sakakibara, Li Wang, Koichi Suto, Chihiro Inoue
ABSTRACT
Fourteen different alkaline coal fly ashes (CFAs) were used for the experiment, in which each sample was mixed with water to be 28.6% of water content (wt/wt) and aged for 1-4 weeks at 10-30 °C. This simple treatment is advantageous for decreases in water-soluble B, F, Cr, and As. Compared to non-aged CFAs, their water-soluble fractions remained 0.56-88%, 21-85%, 0.37-93% and 2.6-88%, respectively, after aging for a week at 20 °C, although the amounts of Cr and As released from some CFA samples increased. Considering the significant decrease in elution of sulfate, Ca and Al after aging, the immobilization, namely prevention of toxic element elution, could be related to formation of secondary minerals such as portlandite, gypsum and ettringite. Immobilization of B and Cr tends to proceed preferentially under colder conditions. Aging at higher temperatures enhances the leachability of Cr in some CFA samples. Contrary to the behavior of B and Cr, water-soluble F effectively decreases under warmer conditions.
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Boron, monofilament, 20m, diameter 0.1mm
Boron, monofilament, 20m, diameter 0.2mm
Boron, monofilament, 10m, diameter 0.1mm
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Chromium, microfoil, disks, 25mm, thinness 0.05μm, specific density 37.27μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, foil, 100x100mm, thickness 3.0mm, hard, 99.95%
Chromium, foil, 25x25mm, thickness 2.0mm, hard, 99.7+%
Chromium, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chromium, bar, 100mm x 2mm x 2mm, 99.7+%
Chromium, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Boron, monofilament, 200m, diameter 0.1mm
Boron, monofilament, 5m, diameter 0.2mm
Boron, monofilament, 10m, diameter 0.2mm
Boron, monofilament, 5m, diameter 0.1mm
Boron, monofilament, 50m, diameter 0.1mm
Chromium, microfoil, disks, 25mm, thinness 1.0μm, specific density 719μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Chromium, rod, 50mm, diameter 5.0mm, 99.7+%
Chromium, microfoil, disks, 10mm, thinness 0.25μm, specific density 178.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%