Merck
  • Lead toxicity to Lemna minor predicted using a metal speciation chemistry approach.
Lead toxicity to Lemna minor predicted using a metal speciation chemistry approach.
Environmental toxicology and chemistry (2014-07-22)
Paula M C Antunes, Nancy J Kreager
ABSTRACT
In the present study, predictive measures for Pb toxicity and Lemna minor were developed from bioassays with 7 surface waters having varied chemistries (0.5-12.5 mg/L dissolved organic carbon, pH of 5.4-8.3, and water hardness of 8-266 mg/L CaCO3 ). As expected based on water quality, 10%, 20%, and 50% inhibitory concentration (IC10, IC20, and IC50, respectively) values expressed as percent net root elongation (%NRE) varied widely (e.g., IC20s ranging from 306 nM to >6920 nM total dissolved Pb), with unbounded values limited by Pb solubility. In considering chemical speciation, %NRE variability was better explained when both Pb hydroxides and the free lead ion were defined as bioavailable (i.e., f{OH} ) and colloidal Fe(III)(OH)3 precipitates were permitted to form and sorb metals (using FeOx as the binding phase). Although cause and effect could not be established because of covariance with alkalinity (p = 0.08), water hardness correlated strongly (r(2)  = 0.998, p < 0.0001) with the concentration of total Pb in true solution ([Pb]T_True solution ). Using these correlations as the basis for predictions (i.e., [Pb]T_True solution vs water hardness and %NRE vs f{OH} ), IC20 and IC50 values produced were within a factor of 2.9 times and 2.2 times those measured, respectively. The results provide much needed effect data for L. minor and highlight the importance of chemical speciation in Pb-based risk assessments for aquatic macrophytes.
Product Description
Lead, foil, not light tested, 50x50mm, thickness 0.004mm, 99.99+%
Lead, foil, 0.5m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 1m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.125mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 4mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 2m coil, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 4mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 2m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 4mm disks, thickness 0.006mm, 99.99+%
Lead, foil, 2m coil, thickness 0.05mm, as rolled, 97% (sn 1.5%, sb 1.5%)
Lead, foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 2m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.02mm, 99.99+%
Lead, wire reel, 1m, diameter 3.0mm, as drawn, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.025mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.008mm, 99.99+%
Lead, wire reel, 0.5m, diameter 3.0mm, as drawn, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.03mm, 99.99+%
Lead, rod, 1000mm, diameter 12.7mm, 99.95%
Lead, foil, not light tested, 100x100mm, thickness 0.04mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)