Merck
  • Rapid molecular screening for multidrug-resistant tuberculosis in a resource-limited region of China.
Rapid molecular screening for multidrug-resistant tuberculosis in a resource-limited region of China.
Tropical medicine & international health : TM & IH (2014-07-22)
Dan Zhang, Beizhong Liu, Yufeng Wang, Yu Pang
ABSTRACT
To investigate the molecular characteristics of MDR and XDR strains circulating in Chongqing, China. The drug target genes conferring for rifampicin (RIF), isoniazid (INH), ethambutol (EMB), ofloxacin (OFLX) and kanamycin (KAN) resistance were screened by DNA sequencing to determine the mutation frequencies in this area. Drug susceptibility of 208 MDR isolates revealed that 132 (63.46%) were resistant to streptomycin (SM), 96 (46.15%) to ethambutol (EMB), 51 (24.52%) to ofloxacin (OFLX), and 26 (12.50%) to kanamycin (KAN); six (2.88%) isolates had XDR profiles. In comparison with the drug susceptibility phenotype, the sensitivity of drug resistance by DNA sequencing was 91.83% for RIF, 87.50% for INH, 66.67% for EMB, 74.51% for OFLX and 53.85% for KAN resistance. 12.50% of EMB- and 1.27% of OFLX-susceptible isolates were harboured genetic mutations in embB and gyrA, respectively. Our findings demonstrate that the hot-spot regions localised in rpoB, katG and inhA genes serve as excellent markers for the corresponding drug resistance, while EMB, OFLX or KAN drug-resistant TB cases may not be identifiable by scanning embB, gyrA, rrs and eis promoter in Chongqing, indicating that further studies on the drug resistance mechanisms of EMB, OFLX and KAN are urgently needed to elucidate the low sensitivity between genomic substitutions and drug-resistant phenotype.
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Supelco
Streptomycin sesquisulfate hydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Ethambutol dihydrochloride, antimycobacterial
Ethambutol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
USP
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Ethambutol for system suitability, European Pharmacopoeia (EP) Reference Standard