Merck
  • Scapular dyskinesis is detrimental to shoulder tendon properties and joint mechanics in a rat model.
Scapular dyskinesis is detrimental to shoulder tendon properties and joint mechanics in a rat model.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2014-07-30)
Katherine E Reuther, Stephen J Thomas, Jennica J Tucker, Sarah M Yannascoli, Adam C Caro, Rameen P Vafa, Stephen S Liu, Joshua A Gordon, Pankti R Bhatt, Andrew F Kuntz, Louis J Soslowsky
ABSTRACT
Shoulder tendon injuries are frequently seen in the presence of abnormal scapular motion, termed scapular dyskinesis. The cause and effect relationship between scapular dyskinesis and shoulder injury has not been directly defined. We developed and used an animal model to examine the initiation and progression of pathological changes in the rotator cuff and biceps tendon. Sixty male Sprague-Dawley rats were randomized into two groups: nerve transection (to induce scapular dyskinesis, SD) or sham nerve transection (control). The animals were euthanized 4 and 8 weeks after surgery. Shoulder function and passive joint mechanics were evaluated over time. Tendon mechanical, histological, organizational, and compositional properties were evaluated at both time points. Gross observation demonstrated alterations in scapular motion, consistent with scapular "winging." Shoulder function, passive internal range of motion, and tendon mechanical properties were significantly altered. Histology results, consistent with tendon pathology (rounded cell shape and increased cell density), were observed, and protein expression of collagen III and decorin was altered. This study presents a new model of scapular dyskinesis that can rigorously evaluate cause and effect relationships in a controlled manner. Our results identify scapular dyskinesis as a causative mechanical mechanism for shoulder tendon pathology.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance